8-Bromoisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

HC-6020-5G HC-6020-1G
COBBHC-6020-5GEA 0 GBP
COBBHC-6020-5G COBBHC-6020-1G
8-Bromoisoquinoline
8-Bromoisoquinoline
Formula: C₉H₆BrN
MDL Number: MFCD04973298
CAS Number: 63927-22-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR