2-Methoxy-N-tert-butylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CA-4792-25G CA-4792-1G CA-4792-5G
COBBCA-4792-25GEA 0 GBP
COBBCA-4792-25G COBBCA-4792-1G COBBCA-4792-5G
2-Methoxy-N-tert-butylbenzamide
2-Methoxy-N-tert-butylbenzamide
Formula: C₁₂H₁₇NO₂
MDL Number: MFCD00751503
CAS Number: 88105-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR