N-Ethyl-3-fluorobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CA-4766-5G
COBBCA-4766-5GEA 0 GBP
COBBCA-4766-5G
N-Ethyl-3-fluorobenzamide
N-Ethyl 3-fluorobenzamide
Formula: C₉H₁₀FNO
MDL Number: MFCD01338769
CAS Number: 150079-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR