Methyl-2-chloronicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methyl 2-chloronicotinate, Methyl 2-chloropyridine-3-carboxylate

CA-4255-5G CA-4255-25G CA-4255-100G
COBBCA-4255-5GEA 0 GBP
COBBCA-4255-5G COBBCA-4255-25G COBBCA-4255-100G
Methyl-2-chloronicotinate
Methyl 2-chloronicotinate
Formula: C₇H₆ClNO₂
MDL Number: MFCD01318629
CAS Number: 40134-18-7
EINECS: 254-805-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR