2-Chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid

BB-2259-5G BB-2259-25G BB-2259-100G
COBBBB-2259-5GEA 0 GBP
COBBBB-2259-5G COBBBB-2259-25G COBBBB-2259-100G
2-Chlorophenylboronic acid
2-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MDL Number: MFCD00674012
CAS Number: 3900-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR