4-(Ethoxycarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

BB-2113-5G BB-2113-25G
COBBBB-2113-5GEA 0 GBP
COBBBB-2113-5G COBBBB-2113-25G
4-(Ethoxycarbonyl)phenylboronic acid
4-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
MDL Number: MFCD02179441
CAS Number: 4334-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR