Horizontal electrophoresis system, multiSUB maxi
Supplier: CLEAVER SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MSMAXI20 MSMAXIDUO MSMAXI25
CLEVMSMAXI20EA 1430 GBP
CLEVMSMAXI20 CLEVMSMAXIDUO CLEVMSMAXI25
Horizontal electrophoresis system, multiSUB maxi
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The multiSUB maxi is designed for resolution of high numbers of samples, such as from cloning and PCR. Tray sizes correspond to standard blotter sizes allows easy sample transfer onto a membrane for further analysis. Two tray options are available: 200×200 mm and 200×250 mm.
  • Run up to 550 samples
  • Low buffer volumes
  • Ideal for extended separations
*Multichannel pipette compatible