Gradient mixers
Supplier: CLEAVER SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CSL-GM15 CSL-GM25 CSL-GM50 CSL-GM100 CSL-GM500
CLEVCSL-GM15EA 1120 GBP
CLEVCSL-GM15 CLEVCSL-GM25 CLEVCSL-GM50 CLEVCSL-GM100 CLEVCSL-GM500
Gradient mixers
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Gradient mixer is ideal for caesium, sucrose and gel gradients.
  • It comprise two chambers-a reservoir and a mixing chamber with an interconnecting valve
  • Second valve regulates the output flow from the mixing chamber
The support rod allows the mixer to be fixed to a retort stand for extra stability.