Laminar flow cabinets, CrossFlow CLF series
Supplier: CleanAir by Baker
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1560401 1560402 1560403 1560404 1560405 1560406
CLEA1560401EA 7650 GBP
CLEA1560401 CLEA1560402 CLEA1560403 CLEA1560404 CLEA1560405 CLEA1560406
Laminar flow cabinets, CrossFlow CLF series
Fume Hoods and Enclosures Laminar Flow Cabinets
The CrossFlow series is a high quality crossflow cabinet range for handling non-hazardous materials that require a clean working environment. The ergonomically designed HEPA filtered horizontal laminar airflow (crossflow) creates an ISO Class 5 (14644-1) / GMP Class A (Annex 1) work area with low noise level and prevents ambient air from entering the work area, ensuring product protection.
  • Microprocessor flow control
  • User-friendly control panel for intuitively operating
  • HEPA filter H14 (efficiency of 99,995% MPPS) and pre-filter (ISO 16890)
  • Energy saving
More Product Information