Colchicine

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

9000760-100 9000760-250 9000760-500
CAYM9000760-100EA 41.4 GBP
CAYM9000760-100 CAYM9000760-250 CAYM9000760-500
Colchicine
Colchicine
Formula: C₂₂H₂₅NO₆
Melting Pt: 154…156 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,I
Merck Index: 12,02536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR