Griess reagent R2
Supplier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
780020-8 780020-50
CAYM780020-8EA 50.8 GBP
CAYM780020-8 CAYM780020-50
Griess reagent R2
Griess reagent
Griess Reagent component for determination of nitrite in aqueous solutions.