Baicalein

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

70610-100 70610-50 70610-500 70610-1
CAYM70610-100EA 80.8 GBP
CAYM70610-100 CAYM70610-50 CAYM70610-500 CAYM70610-1
Baicalein
Baicalein
Formula: C₁₅H₁₀O₅
MDL Number: MFCD00017459
CAS Number: 491-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR