13,14-dihydro-15-keto Prostaglandin F₂α ELISA kit
Supplier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
516671-480 516671-96 516671-96S 516671-480S
CAYM516671-480EA 2980 GBP
CAYM516671-480 CAYM516671-96 CAYM516671-96S CAYM516671-480S
13,14-dihydro-15-keto Prostaglandin F₂α ELISA kit
Assays ELISAs
This kit is used for the measurement of 13,14-dihydro-15-keto prostaglandin F₂α in plasma.
  • Detect 13,14-dihydro-15-keto PGF₂α, a stable metabolite of PGF₂α
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 13,14-dihydro-15-keto PGF₂α levels down to 16,3 pg/ml
  • Reads colorimetric at 405 to 420 nm
PGF₂α is rapidly metabolized to 13,14-dihydro-15-keto PGF₂α in vivo, by the enzymes 15-hydroxy PG dehydrogenase and D13-reductase. Measurement of 13,14-dihydro-15-keto PGF₂α in plasma can be used as a marker of the in vivo production of PGF₂α.
13,14-dihydro-15-keto PGF₂α ELISA is a competitive assay that provides measurements of 13,14-dihydro-15-keto PGF₂α within the range of 2,3 to 5000 pg/ml. The assay has been validated for the measurement of 13,14-dihydro-15-keto PGF₂α in plasma, a common matrix for measurement of this PGF₂α metabolite.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.