Riboflavin 5'-monophosphate sodium salt hydrate
Supplier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18167-10 18167-100 18167-25 18167-50
CAYM18167-10EA 82.7 GBP
CAYM18167-10 CAYM18167-100 CAYM18167-25 CAYM18167-50
Riboflavin 5'-monophosphate sodium salt hydrate
Enzymes
Riboflavin 5'-monophosphate (FMN) is a coenzyme that is tightly bound to enzymes catalysing oxidation and reduction reactions in a variety of biosynthetic pathways.
  • Soluble in organic solvents - ethanol; DMSO and DMF
  • Solubility in PBS is 10 mg/ml
FMN also binds the FMN riboswitch (RFN element) on RNA to alter gene regulation. FMN is a substrate of FMN phosphohydrolases and is used to study their function.
Synonyms: Riboflavin 5'-(dihydrogen phosphate), monosodium salt, dihydrate; flavin mononucleotide, FMN, vitamin B2 phosphate.