7-Ethoxyresorufin ≥95%, solid

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

16122-10 16122-1
CAYM16122-10EA 527 GBP
CAYM16122-10 CAYM16122-1
7-Ethoxyresorufin ≥95%, solid
7-Ethoxyresorufin

7-Ethoxyresorufin (7-ER) is a fluorometric substrate for and competitive inhibitor of cytochrome P450 1A1 (Km ranges from 0,11-0,23 µM; IC50 = 0,1 µM)


It has been used to study the metabolism of arachidonic acid and the mechanisms of vasodilation. 7-ER also has been shown to inhibit nitric oxide synthesis by uncoupling neuronal nitric oxide synthase (Ki = 0,76 µM)

Formula: C₁₄H₁₁NO₃
MDL Number: MFCD00037661
CAS Number: 5725-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR