Anti-ALDH1 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

160720-1
CAYM160720-1EA 786 GBP
CAYM160720-1
Anti-ALDH1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDH1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Sheep, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR