Acridine orange

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14338-5 14338-10 14338-25
CAYM14338-5EA 156 GBP
CAYM14338-5 CAYM14338-10 CAYM14338-25
Acridine orange
Acridine orange
Formula: C₁₇H₁₉N₃
MDL Number: MFCD00005029
CAS Number: 494-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR