Fast green FCF

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt

14335-5 14335-10 14335-25 14335-100
CAYM14335-5EA 95.9 GBP
CAYM14335-5 CAYM14335-10 CAYM14335-25 CAYM14335-100
Fast green FCF
Fast Green FCF
Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR