Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13491-1
CAYM13491-1EA 1000 GBP
CAYM13491-1
Anti-HDAC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HDAC1
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
Immunogen synthetic peptides corresponding to amino acids 1-5, 433-448 and 467-482 of human HDAC1
Amino acid number 1 to 5 433 to 448, 467 to 482
Purification purified by Protein G chromatography
Cross adsorption No
Storage buffer PBS containing 0,02% sodium azide
Storage temperature –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR