G418 disulphate

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

13200-1 13200-5
CAYM13200-1EA 141 GBP
CAYM13200-1 CAYM13200-5
G418 disulphate
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR