2-Methoxyestradiol

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

13021-5 13021-10 13021-25 13021-50
CAYM13021-5EA 79 GBP
CAYM13021-5 CAYM13021-10 CAYM13021-25 CAYM13021-50
2-Methoxyestradiol
2-Methoxyestradiol
Formula: C₁₉H₂₆O₃
MW: 302.408 g/mol
MDL Number: MFCD00010489
CAS Number: 362-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR