Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13007-100 13007-250 13007-500 13007-1
CAYM13007-100EA 41.4 GBP
CAYM13007-100 CAYM13007-250 CAYM13007-500 CAYM13007-1
Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Formula: C₁₂H₁₁BrN₂O₂S
MDL Number: MFCD11869291
CAS Number: 1138444-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR