Indole-4-carbaldehyd

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10007086-1 10007086-10 10007086-5 10007086-500
CAYM10007086-1EA 356 GBP
CAYM10007086-1 CAYM10007086-10 CAYM10007086-5 CAYM10007086-500
Indole-4-carbaldehyd
Indole-4-carbaldehyd
Formula: C₉H₇NO
MDL Number: MFCD01632221
CAS Number: 1074-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR