Human aortic smooth muscle cells (HAoSMC)
Supplier: PROMOCELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C-12533 C-14053
C-12533EA 1160 GBP
C-12533 C-14053
Human aortic smooth muscle cells (HAoSMC)
Cells
Primary human aortic smooth muscle cells (HAoSMC) are isolated from plaque-free regions of the human aorta and stain positive for smooth muscle α-actin.
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
The pulsatile pressure produced by the heart causes a cyclic distention of the aorta. The smooth muscle cells in the aortic wall subsequently contract it again. Changes in the arterial wall, associated with vascular diseases such as atherosclerosis and hypertension, strongly influence this process.
HAoSMC are suitable for studying the role of smooth muscle cells under normal or disease conditions in vitro.
Ordering information: Human Aortic Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
SPECIFICATIONS
Environmentally Preferable