Multi-gas detector, GasAlert Max XT II
Supplier: BW TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
XT-0W0M-Y-EU XT-X000-Y-EU XT-XW00-Y-EU XT-XW0M-Y-EU XT-XWHM-Y-EU XT-0W00-Y-EU XT-X00M-Y-EU XT-0W00-Y-UK XT-0W0M-Y-UK XT-X000-Y-UK XT-X00M-Y-UK XT-XW00-Y-UK XT-XW0M-Y-UK XT-XWHM-Y-UK
BWTEXT-0W0M-Y-EUEA 688 GBP
BWTEXT-0W0M-Y-EU BWTEXT-X000-Y-EU BWTEXT-XW00-Y-EU BWTEXT-XW0M-Y-EU 151-0239 BWTEXT-0W00-Y-EU BWTEXT-X00M-Y-EU BWTEXT-0W00-Y-UK BWTEXT-0W0M-Y-UK BWTEXT-X000-Y-UK BWTEXT-X00M-Y-UK BWTEXT-XW00-Y-UK BWTEXT-XW0M-Y-UK BWTEXT-XWHM-Y-UK
Multi-gas detector, GasAlert Max XT II
Gas Detection Systems
The rugged GasAlert Max XT II reliably monitors up to four gas hazards and combines straightforward one-button field operation with an integrated sampling pump.
  • Compatible with BW MicroDock II automatic test and calibration station
  • Powered by a rechargeable battery pack
  • GasAlert Max XT II comes with standard event logging and datalogging (accessory connectivity kit required)
Tamper-proof, user-adjustable options enable the instrument to be customised to suit your application.
Delivery information: Supplied with detector complete with sensors,More Product Information