Multi-gas detector, GasAlert Micro 5
Supplier: BW TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M5X0AYRDDYU00 M5X0GMRDDYU00 M5XU0YRPDYU00 M5XW00RPDYU00 M5XW0YRDDYU00 M5XW0YRPDYU00 M5XWAMRDDYU00 M5XWAYRDDYU00 M5XWAYRPDYU00 M5XWCYRDDYU00 M5XWCYRPDYU00 M5XWH0RDDYU00 M5XWH0RPDYU00 M5XWM0RDDYU00 M5XWSYRDDYU00 M5XWD0RDDYE00
BWTEM5X0AYRDDYU00EA 0 GBP
BWTEM5X0AYRDDYU00 BWTEM5X0GMRDDYU00 BWTEM5XU0YRPDYU00 BWTEM5XW00RPDYU00 BWTEM5XW0YRDDYU00 BWTEM5XW0YRPDYU00 BWTEM5XWAMRDDYU00 BWTEM5XWAYRDDYU00 BWTEM5XWAYRPDYU00 BWTEM5XWCYRDDYU00 BWTEM5XWCYRPDYU00 BWTEM5XWH0RDDYU00 BWTEM5XWH0RPDYU00 BWTEM5XWM0RDDYU00 BWTEM5XWSYRDDYU00 BWTEM5XWD0RDDYE00
Multi-gas detector, GasAlert Micro 5
Gas Detection Systems
GasAlert Micro 5 was designed for protecting five potential atmospheric hazards including oxygen, combustible and toxic gases.
  • Integral concussion-proof boot
  • Four alarm levels: Low, High, TWA and STEL
  • Compatible with MicroDock II automatic test and calibration station
It continuously monitors and displays concentrations of up to five gases.
Delivery information: Supplied with a detector complete with specified sensor(s), stainless steel alligator clip, concussion-proof housing, cradle charger, wall outlet charging adaptor, screwdriver, calibration/More Product Information