Anti-FUT2 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS7813
BWRLBS7813EA 408 GBP
BWRLBS7813
Anti-FUT2 Rabbit Polyclonal Antibody
Antibodies
Recombinant full length Human FUT2.

FUT2 pAb detects endogenous levels of FUT2 protein.

Type: Primary
Antigen: FUT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR