Anti-SLC3A1 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS7609
BWRLBS7609EA 408 GBP
BWRLBS7609
Anti-SLC3A1 Rabbit Polyclonal Antibody
Antibodies
Recombinant full length Human SLC3A1.

SLC3A1 pAb detects endogenous levels of SLC3A1 protein.

Type: Primary
Antigen: SLC3A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR