Anti-MBP Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS6660
BWRLBS6660EA 408 GBP
BWRLBS6660
Anti-MBP Rabbit Polyclonal Antibody
Antibodies
Recombinant full length Human MBP.

MBP pAb detects endogenous levels of MBP protein.

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR