Anti-GPR81 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS60353
BWRLBS60353EA 313 GBP
BWRLBS60353
Anti-GPR81 Rabbit Polyclonal Antibody
Antibodies
A synthetic peptide corresponding to residues in Human GPR81.

GPR81 pAb detects endogenous levels of GPR81 protein.

Type: Primary
Antigen: GPR81
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR