Anti-USP19 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS5975
BWRLBS5975EA 313 GBP
BWRLBS5975
Anti-USP19 Rabbit Polyclonal Antibody
Antibodies
Synthetic peptide, corresponding to amino acids 439-484 of Human USP19.

USP19 pAb detects endogenous levels of USP19 protein.

Type: Primary
Antigen: USP19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR