Anti-TFEB Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS5963
BWRLBS5963EA 313 GBP
BWRLBS5963
Anti-TFEB Rabbit Polyclonal Antibody
Antibodies
Synthetic peptide, corresponding to amino acids 6-51 of Human TFEB.

  • TFEB pAb detects endogenous levels of TFEB protein.

Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR