Anti-RAPGEF5 Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS5890
BWRLBS5890EA 313 GBP
BWRLBS5890
Anti-RAPGEF5 Rabbit Polyclonal Antibody
Antibodies
Synthetic peptide, corresponding to amino acids 476-521 of Human RAPGEF5.

RAPGEF5 pAb detects endogenous levels of RAPGEF5 protein.

Type: Primary
Antigen: RAPGEF5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR