Anti-SHIP1 Rabbit Polyclonal Antibody [clone: M1014]

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS1595
BWRLBS1595EA 313 GBP
BWRLBS1595
Anti-SHIP1 Rabbit Polyclonal Antibody [clone: M1014]
Antibodies
Synthetic peptide, corresponding to amino acids 980-1030 of Human SHIP1.

SHIP1 (M1014) pAb detects endogenous levels of SHIP1 protein.

Type: Primary
Antigen: SHIP1
Clonality: Polyclonal
Clone: M1014
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR