Anti-PKC delta Rabbit Polyclonal Antibody [clone: G499]

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS1578
BWRLBS1578EA 313 GBP
BWRLBS1578
Anti-PKC delta Rabbit Polyclonal Antibody [clone: G499]
Antibodies
Synthetic peptide, corresponding to amino acids 470-520 of Human PKC delta.

PKC ? (G499) pAb detects endogenous levels of PKC ? protein.

Type: Primary
Antigen: PKC delta
Clonality: Polyclonal
Clone: G499
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

SPECIFICATIONS

Antigen symbol PKC delta
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Clone G499
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR