Anti-Raf-B Rabbit Polyclonal Antibody [clone: F594]

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS1515
BWRLBS1515EA 313 GBP
BWRLBS1515
Anti-Raf-B Rabbit Polyclonal Antibody [clone: F594]
Antibodies
Synthetic peptide, corresponding to amino acids 560-610 of Human Raf-B.

B-Raf (F594) pAb detects endogenous levels of B-Raf protein.

Type: Primary
Antigen: Raf-B
Clonality: Polyclonal
Clone: F594
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR