Anti-SRF Rabbit Polyclonal Antibody [clone: G93]

Supplier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS1332
BWRLBS1332EA 313 GBP
BWRLBS1332
Anti-SRF Rabbit Polyclonal Antibody [clone: G93]
Antibodies
Synthetic peptide, corresponding to amino acids 61-110 of Human SRF.

SRF (G93) pAb detects endogenous levels of SRF protein.

Type: Primary
Antigen: SRF
Clonality: Polyclonal
Clone: G93
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR