Pipette container and basket
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9650-0653 9650-0652 9650-0651
BURK9650-0653EA 82.7 GBP
BURK9650-0653 BURK9650-0652 BURK9650-0651
Pipette container and basket
Baskets Pipette Baskets
Made of PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
For cleaning and storage of pipettes. Containers and basket may be used as accessories for the pipette rinsing system.