Pipette rinsing system
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9650-0650
BURK9650-0650EA 425 GBP
BURK9650-0650
Pipette rinsing system
Pipettes Pipette Washers
For easy and gentle cleaning and safe storage of pipettes. A complete system suitable for every lab.
  • PE transparent
  • Effective height of 600 mm
The basket is an accessory for the pipette rinser.