Syringes, SteriPlast®
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5325-0100 5325-0060
BURK5325-0100EA 112 GBP
BURK5325-0100 BURK5325-0060
Syringes, SteriPlast®
Syringes Syringes, Medical
Transparent PP syringes with printed graduations.
  • Can be extended to any length with additional tubing
  • Luer adapter is supplied for the 100 ml syringe
The syringe is supplied in a sterile package and ready for immediate use.