Tanks, rectangular, LDPE, LaboPlast®
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3440-0231 3440-0235 3440-1551 3440-1556 3440-2506 3440-2501 3440-1906 3440-3207 3440-4007 3440-2857 3440-2607 3440-0505 3440-1156 3440-1006 3440-1186 3440-1506 3440-0485 3440-0855 3440-0135 3440-0455 3440-0481 3440-0901 3440-0906 3440-1301 3440-1306 3440-1001 3440-0131 3440-3201 3440-0851 3440-0501 3440-4001
BURK3440-0231EA 319 GBP
BURK3440-0231 BURK3440-0235 BURK3440-1551 BURK3440-1556 BURK3440-2506 BURK3440-2501 BURK3440-1906 BURK3440-3207 BURK3440-4007 BURK3440-2857 BURK3440-2607 BURK3440-0505 BURK3440-1156 BURK3440-1006 BURK3440-1186 BURK3440-1506 BURK3440-0485 BURK3440-0855 BURK3440-0135 BURK3440-0455 BURK3440-0481 BURK3440-0901 BURK3440-0906 BURK3440-1301 BURK3440-1306 BURK3440-1001 BURK3440-0131 BURK3440-3201 BURK3440-0851 BURK3440-0501 BURK3440-4001
Tanks, rectangular, LDPE, LaboPlast®
Tanks Rectangular Tanks
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
  • Linear LDPE, transparent
  • Rupture-proof