Bottles, aluminium, UN approved, with screw cap
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0329-0125 0329-1250 0329-0500 0329-0250
BURK0329-0125EA 33.8 GBP
BURK0329-0125 BURK0329-1250 BURK0329-0500 BURK0329-0250
Bottles, aluminium, UN approved, with screw cap
Bottles Metal Bottles
Pure aluminium (material AL 99,5).
  • UN code is printed onto the bottles in black
  • With eyelets, sealable (except the 125 ml model)
  • Light, unbreakable, seamless
Aluminium transport bottles with screw cap close safely and tightly and are approved for packaging type I (X).