Stainless steel cleaner, Suma Inox D7.1, Diversey
Supplier: BUNZL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
042504
BUNZ042504EA 0 GBP
BUNZ042504
Stainless steel cleaner, Suma Inox D7.1, Diversey
Cleaning Agents Detergents and Surface Cleansers
Suma Inox D7.1 is a ready to use one step light duty cleaner/polish for use on stainless steel surfaces that are not used in the preparation of food. It gives shine back to stainless steel surfaces, removes spots and marks, and resists finger marks to maintain the just polished appearance for longer.