Electroporation system, high throughput ECM™ 830
Supplier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45-0665 45-0666
BTXI45-0665EA 15470 GBP
BTXI45-0665 BTXI45-0666
Electroporation system, high throughput ECM™ 830
Electroporation Systems Electroporators
The High Throughput ECM™ 830 system offers a multi-well electroporation technology for processing multiple samples in seconds. Using the HT multi-well plates instead of traditional cuvettes, the researcher can transition from a single cuvette to either a 25-well or 96-well electroporation plate by using one simple plate handler. This increases yields and the number of experiments run in a single day. Experiments take seconds to run, allowing for quick and efficient optimisation of the electrical and biological parameters. Once optimised, samples are rapidly processed increasing yields andMore Product Information