Platinum electrodes, Tweezertrodes™
Supplier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45-0486 45-0487 45-0488 45-0489
BTXI45-0486EA 1550 GBP
BTXI45-0486 BTXI45-0487 BTXI45-0488 BTXI45-0489
Platinum electrodes, Tweezertrodes™
Electrodes Electroporation Electrodes
Tweezertrodes™ are reusable, tweezer style electrodes for in vivo, and in utero drug or gene delivery. Tweezertrodes™ consist of a standard 12 cm tweezer that has been modified with stainless steel or platinum circular electrodes at the tip. The gap between the electrode disks may be adjusted from under 1 mm to over 2 cm.
  • May be cleaned with a mild detergent and sterilised with ethanol or ethylene oxide
  • Can be connected to an electroporator via a connection cable
  • Compatible with the BTX ECM® 630, 830 and 2001
  • Ideal for in vivo and in utero applications in animal tissues
Order Now
SPECIFICATIONS
Environmentally Preferable