Anti-dephosphorylated Catenin beta Sheep Polyclonal Antibody
Supplier: Biosensis
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S-1233-100
BSENS-1233-100EA 359 GBP
BSENS-1233-100
Anti-dephosphorylated Catenin beta Sheep Polyclonal Antibody
Antibodies
Catenin beta is an adherens junction protein and has a role in the regulation of cell adhesion and in signal transduction through the Wnt pathway. At least 2 isoforms are produced by alternative splicing.
The specificity of this antibody has been confirmed by WB against the antigen.
Application Information:
Western Blotting (WB) and Immunocytochemistry. A concentration of 2.0 µg/ml is recommended for WB. Human Catenin beta (isoform 1) has a predicted length of 781 residues and MW of 86 kDa. It has been observed as a 92 kDa band in SDS-reducing gels. A concentration of 4.0 µg/ml isMore Product Information