Anti-alpha synuclein Rabbit Polyclonal Antibody
Supplier: Biosensis
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R-075-100
BSENR-075-100EA 304 GBP
BSENR-075-100
Anti-alpha synuclein Rabbit Polyclonal Antibody
Antibodies
Alpha synuclein is an abundant 140 amino acid neuronal protein, expressed primarily at presynaptic terminals in the central nervous system. FUNCTION: May be involved in the regulation of dopamine release and transport. Soluble protein, normally localized primarily at the presynaptic region of axons, which can form filamentous aggregates that are the major non amyloid component of intracellular inclusions in several neurodegenerative diseases (synucleinopathies). Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leadingMore Product Information