TZe Labelling tape cartridges for P-Touch
Supplier: BROTHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TZE-441 TZE-325 TZE-721 TZE-222 TZE-211 TZE-551 TZE-252 TZE-354 TZE-111 Z243 TZE-242 TZE-335 TZE-232 TZE-345 TZE-223 TZE-151 TZE-141 TZE-132 TZE-131 TZE-121 TZE-233 TZE-C31 TZE-B31 TZE-741 TZE-435 TZE-421 TZE611 TZE-135 TZE-S231
811-0888EA 0 GBP
811-0888 811-0879 811-0901 811-0866 811-0864 811-0894 811-0875 811-0884 BROTTZE-111 BROTZ243 BROTTZE-242 BROTTZE-335 BROTTZE-232 BROTTZE-345 BROTTZE-223 BROTTZE-151 BROTTZE-141 BROTTZE-132 BROTTZE-131 BROTTZE-121 BROTTZE-233 BROTTZE-C31 BROTTZE-B31 BROTTZE-741 BROTTZE-435 BROTTZE-421 BROTTZE611 BROTTZE-135 BROTTZE-S231
TZe Labelling tape cartridges for P-Touch
Labels
These TZe labelling tapes are developed for use with P-Touch labelprinters. They feature the exclusive laminated tape process and standard adhesive. Best suited for smooth flat surfaces.
  • Perfect for everyday applications
  • Stay on under normal indoor and outdoor use, including hot and cold environments
Length: 8,0 m
Different colours available.