Burettes, straight stopcock, Schellbach, with USP individual or DAkkS calibration certificate, BLAUBRAND®
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DAKKS12484 USP12484 USP12487 USP12488 DAKKS12488
BRNDDAKKS12484EA 169 GBP
BRNDDAKKS12484 BRNDUSP12484 BRNDUSP12487 BRNDUSP12488 BRNDDAKKS12488
Burettes, straight stopcock, Schellbach, with USP individual or DAkkS calibration certificate, BLAUBRAND®
Burettes
Borosilicate glass 3.3, class AS.
  • Calibrated to deliver 'TD, Ex', conformity certified
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm