Bulb pipettes (one mark), with individual certificate, BLAUBRAND®, Class AS
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
929712 929714 929717 929707 929702 929703 929705 929709 929718 929719 929704 929713 929716 929706 929708 929710 929711 929715
612-1074EA 63 GBP
612-1074 BRND929714 BRND929717 BRND929707 BRND929702 BRND929703 BRND929705 BRND929709 BRND929718 BRND929719 BRND929704 BRND929713 BRND929716 BRND929706 BRND929708 BRND929710 BRND929711 BRND929715
Bulb pipettes (one mark), with individual certificate, BLAUBRAND®, Class AS
Pipettes Bulb Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • Also available as separate USP version
  • Batch number, individual serial number, measured volume, measurement uncertainty, and date of issue are documented on the certificate
Certifications: DIN EN ISO 648; DE-M marked.